2-acetyl-5-bromo-4,7-dihydro-1-benzofuran-4,7-dione ≥97%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2,2',2'',2'''-(2-(4-Isothiocyanatobenzyl)-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetic acid

A173817-250MG
ALA173817250MGEA 1442.96 USD
ALA173817250MG
2-acetyl-5-bromo-4,7-dihydro-1-benzofuran-4,7-dione ≥97%
2-acetyl-5-bromo-4,7-dihydro-1-benzofuran-4,7-dione
Formula: C₁₀H₅BrO₄
Storage Temperature: Ambient
CAS Number: 1389264-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=551a778d72e8f56e6f9db2b332b9f658820208be&vid=551a778d72e8f56e6f9db2b332b9f658820208be&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR