(A-amino-5-bromopyridin-2-yl)methanol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A173629-250MG
ALA173629250MGEA 1442.96 USD
ALA173629250MG
(A-amino-5-bromopyridin-2-yl)methanol ≥97%
(A-amino-5-bromopyridin-2-yl)methanol
Formula: C₆H₇BrN₂O
Storage Temperature: Ambient
CAS Number: 1363381-68-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23d02861d23fe5c55b94ba1faf52affff35106a4&vid=23d02861d23fe5c55b94ba1faf52affff35106a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR