1-Allyl-3-methylimidazolium bromide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A169503-100G A169503-1G A169503-25G A169503-5G
ALA169503-100GEA 367.56 USD
ALA169503-100G ALA169503-1G ALA169503-25G ALA169503-5G
1-Allyl-3-methylimidazolium bromide ≥97%
1-Allyl-3-methylimidazolium bromide

1-Allyl-3-methylimidazolium bromide can be used to prepare:· A weak gel of chitin.· Composite gels of chitin/cellulose.· Methylimidazolium based ionic liquids with varying anions as electrolytes in graphene nanosheet-based supercapacitors.

Formula: C₇H₁₁N₂Br
Melting Pt: 54.0—58.0°C
Storage Temperature: Ambient
MDL Number: MFCD08277003
CAS Number: 31410-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6a38141d21675cf9ab0de037e10b4d380bc538a&vid=a6a38141d21675cf9ab0de037e10b4d380bc538a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR