2-Amino-5-chlorobenzonitrile ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Chloroanthranilonitrile

A151558-1G A151558-25G A151558-5G A151558-100G
ALA151558-1GEA 10.36 USD
ALA151558-1G ALA151558-25G ALA151558-5G ALA151558-100G
2-Amino-5-chlorobenzonitrile ≥98% (by GC)
2-Amino-5-chlorobenzonitrile

The vibrational spectra of 2-amino-5-chlorobenzonitrile was studied using density functinal theory.2-Amino-5-chlorobenzonitrile was used to build up the 2,2-dioxide-1H-2,1,3-benzothiadiazine ring system.

Formula: C₇H₅ClN₂
Boiling Pt: 159 °C
Melting Pt: 96 °C
MDL Number: MFCD00017106
CAS Number: 5922-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0351032382dc49e30f8179a4a08e1ef21a79de9&vid=c0351032382dc49e30f8179a4a08e1ef21a79de9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR