1-(3-Aminopropyl)imidazole ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: N-(3-Aminopropyl)-imidazole, Imidazole-1-propylamine, 1-(3-Aminopropyl)-1H-imidazole

A151397-50G A151397-10G A151397-250G
ALA151397-50GEA 53.11 USD
ALA151397-50G ALA151397-10G ALA151397-250G
1-(3-Aminopropyl)imidazole ≥97% (by GC)
1-(3-Aminopropyl)imidazole

It was used in the synthesis of pH-sensitive polyaspartamide derivatives. It was also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

Formula: C₆H₁₁N₃
Boiling Pt: 117°C
Flash Pt: 154°C
Storage Temperature: Ambient
MDL Number: MFCD00009819
CAS Number: 5036-48-6
EINECS: 225-730-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8139871decbba68f87c2d34d64778a96d7ace3f1&vid=8139871decbba68f87c2d34d64778a96d7ace3f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR