4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 4-Amino-6-hydroxy-2-mercaptopyrimidine Hydrate, 6-Amino-2-thiouracil hydrate, 6-Amino-2-thiouracil Hydrate

A151319-25G
ALA151319-25GEA 21.76 USD
ALA151319-25G
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98%
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate

4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.

Formula: C₄H₅N₃OS·xH₂O
Melting Pt: >300°C
MDL Number: MFCD00052384
CAS Number: 65802-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ca2bd01377a832473d5c1c118d875241ff294db&vid=7ca2bd01377a832473d5c1c118d875241ff294db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR