2,4-Diaminophenol dihydrochloride ≥98% (by total nitrogen and titration analysis)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol

A151245-100G A151245-1G A151245-25G A151245-5G
ALA151245-100GEA 113.91 USD
ALA151245-100G ALA151245-1G ALA151245-25G ALA151245-5G
2,4-Diaminophenol dihydrochloride ≥98% (by total nitrogen and titration analysis)
2,4-Diaminophenol dihydrochloride
Formula: C₆H₁₀Cl₂N₂O
Melting Pt: 205°C
MDL Number: MFCD00012979
CAS Number: 137-09-7
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae7858110a4d021ddab64c40044511b2fcc71c6&vid=2ae7858110a4d021ddab64c40044511b2fcc71c6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR