2-Amino-1-phenylethanol ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: alpha-(Aminomethyl)benzyl Alcohol, DL-β-Hydroxyphenethylamine, Phenylethanolamine

A151123-1G A151123-25G A151123-5G
ALA151123-1GEA 46.46 USD
ALA151123-1G ALA151123-25G ALA151123-5G
2-Amino-1-phenylethanol ≥97% (by GC)
2-Amino-1-phenylethanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008137
CAS Number: 7568-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ffe0c989eac130c04e922fc61ba134e667741ae&vid=2ffe0c989eac130c04e922fc61ba134e667741ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR