α,α'-Dibromo-m-xylene ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

A133958-100G A133958-5G
ALA133958-100GEA 570.86 USD
ALA133958-100G ALA133958-5G
α,α'-Dibromo-m-xylene ≥97% (by GC)
α,α'-Dibromo-m-xylene

Product Introduction:α,α′-Dibromo-m-xylene has been used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macrocyclic salt.

Formula: C₈H₈Br₂
Boiling Pt: 140°C
Melting Pt: 74-76°C
MDL Number: MFCD00000178
CAS Number: 626-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f85c0a5a251529c80d98ed1c7cf5cda1a3e0ba66&vid=f85c0a5a251529c80d98ed1c7cf5cda1a3e0ba66&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR