Acyclovir ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC

Synonyms: 1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on, Acycloguanosine, 2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one, Acyclovir, 9-[(2-Hydroxyethoxy)methyl]guanine, 2-Amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6H-purin-6-one, 2-Amino-1,9-dihydro-9-[(2-hydroxyethoxy)methyl]-6H-purin-6-one, Aciclovir, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one

A126073-100G A126073-1G A126073-25G A126073-5G
ALA126073-100GEA 1348.91 USD
ALA126073-100G ALA126073-1G ALA126073-25G ALA126073-5G
Acyclovir ≥99%, Moligand™
Acyclovir

Aciclovir is a synthetic nucleoside analogue active against herpesviruses.An antiviral compound that can be used to induce apoptosis.

Formula: C₈H₁₁N₅O₃
Melting Pt: 262°C
Storage Temperature: Freezer
MDL Number: MFCD00057880
CAS Number: 59277-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfe6cffd33d47e346044716318d950f963bd240c&vid=dfe6cffd33d47e346044716318d950f963bd240c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR