Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
01090-87306 5021-1816 5021-1817 5021-1823 5067-4656 G1312-67305 G1315-87311 G1316-87300 01090-87611 5021-1818 5021-1819 5021-1820 5021-1821 5021-1822 5022-2159 5065-9931 5065-9963 5067-4655 5067-4659 5067-4669 5500-1157 5500-1217 5500-1243 5500-1246 G1312-67302 G1312-87301 G1312-87303 G1312-87304 G1313-87301 G1313-87305 G1316-87321 G1329-87300 G1367-87303 01048-87302 01078-87305 01090-87304 01090-87308 01090-87610 5021-1831 5065-9932 5065-9933 5065-9935 5065-9964 5067-4637 5067-4647 5067-4648 5067-4649 5067-4650 5067-4651 5067-4652 5067-4653 5067-4657 5067-4658 5067-4660 5067-4670 5067-4685 5067-4688 5067-4721 5067-4722 5067-4723 5067-4724 5067-4735 5067-4744 5067-4745 5067-4746 5067-4748 5067-5104 5067-5107 5067-5108 5067-5111 5067-5113 5067-5416 5067-6129 5500-1199 5500-1200 5500-1201 5500-1202 5500-1203 5500-1238 5500-1240 5500-1242 5500-1245 5500-1249 5500-1250 5500-1251 5500-1252 5500-1253 5500-1272 79841-87610 G1311-81600 G1311-81601 G1312-67300 G1312-67301 G1312-67500 G1312-87300 G1312-87305 G1312-87306 G1313-87304 G1313-87307 G1315-87303 G1315-87312 G1316-27301 G1316-87305 G1316-87309 G1316-87312 G1316-87315 G1316-87316 G1316-87317 G1316-87318 G1316-87319 G1316-87323 G1316-87328 G1328-87600 G1367-87304 G1375-87317 G4220-87000 G4226-60301 G5667-81001 G5667-81003 G5667-81004 G5667-81005 5022-6510 5023-2254 5065-9927 5065-9930 5065-9979 5065-9980 5067-1570 5067-1571 5067-4604 5067-4606 5067-4607 5067-4608 5067-4609 5067-4684 5067-4686 5067-4687 5067-4689 5067-4719 5067-4737 5067-4783 5067-5106 5067-5109 5067-5110 5067-5112 5067-5120 5067-5749 5067-5750 5067-5926 5067-6188 5067-6645 5067-6646 5067-6647 5067-6821 5500-1204 5500-1209 5500-1210 5500-1264 5500-1270 5500-1286 5500-1301 5500-1305 5500-1306 5500-1308 5500-1315 5500-1319 5500-1346 5500-1347 5500-1376 79826-87606 79826-87607 79835-87638 79841-87609 79846-87605 G1313-87308 G1315-87307 G1316-87304 G1316-87306 G1316-87313 G1316-87314 G1316-87326 G1361-67300 G1361-67301 G1361-67302 G1367-60301 G1367-87302 G1375-87304 G1375-87306 G1375-87311 G1375-87312 G1375-87318 G1375-87320 G2258-87301 G2260-87300 G2260-87301 G5611-60500 G5611-60501 G5611-60502 G5611-60503 G5667-81000 G5667-81002 G5667-81007 5004-0016 5004-0017 5004-0018 5004-0019 5004-0041 5022-6508 5022-6509 5061-3361 5065-4493 5065-9906 5065-9926 5067-1572 5067-4605 5067-4784 5067-5709 5067-5713 5067-5939 5067-5941 5067-6643 5067-6644 5067-6820 5500-1278 5500-1280 5500-1284 5500-1300 5500-1302 5500-1303 5500-1309 5500-1314 5500-1316 5500-1317 5500-1318 5500-1320 5500-1329 5500-1330 5500-1333 5500-1334 5500-1335 5500-1336 5500-1384 5500-1385 5500-1386 5500-1387 5500-1388 5500-1390 5500-1391 5500-1393 5500-1398 5500-1406 5500-1407 5500-1408 5500-1415 5500-1423 5500-1432 5500-1433 5500-1434 5500-1438 5500-1441 5500-1442 5500-1444 5500-1445 5500-1447 5500-1451 5500-1456 5500-1462 79826-87601 79826-87605 79826-87608 G1313-87302 G1313-87306 G1315-68708 G1315-87305 G1315-87318 G1315-87323 G1315-87328 G1315-87333 G1315-87338 G1316-87327 G1361-67304 G1361-67305 G1361-67306 G1361-67500 G1361-67501 G1373-87300 G1375-87300 G1375-87301 G1375-87302 G1375-87303 G1375-87305 G1375-87308 G1375-87309 G1375-87310 G1375-87313 G1375-87314 G1375-87316 G1375-87319 G1375-87321 G1375-87322 G1375-87323 G1375-87324 G1375-87325 G1375-87327 G1375-87400 G1377-87301 G4240-87300 G4240-87301 G4240-87302 G4240-87303 G4240-87304 G4240-87310 G4280-81300 G4280-81301 G4301-60056 G5611-67300 G5611-67301 G5615-87302 G7132-60002
AG01090-87306EA 81.3 USD
AG01090-87306 AG5021-1816 AG5021-1817 AG5021-1823 AG5067-4656 AGG1312-67305 AGG1315-87311 AGG1316-87300 AG01090-87611 AG5021-1818 AG5021-1819 AG5021-1820 AG5021-1821 AG5021-1822 AG5022-2159 AG5065-9931 AG5065-9963 AG5067-4655 AG5067-4659 AG5067-4669 AG5500-1157 AG5500-1217 AG5500-1243 AG5500-1246 AGG1312-67302 AGG1312-87301 AGG1312-87303 AGG1312-87304 AGG1313-87301 AGG1313-87305 AGG1316-87321 AGG1329-87300 AGG1367-87303 AG01048-87302 AG01078-87305 AG01090-87304 AG01090-87308 AG01090-87610 AG5021-1831 AG5065-9932 AG5065-9933 AG5065-9935 AG5065-9964 AG5067-4637 AG5067-4647 AG5067-4648 AG5067-4649 AG5067-4650 AG5067-4651 AG5067-4652 AG5067-4653 AG5067-4657 AG5067-4658 AG5067-4660 AG5067-4670 AG5067-4685 AG5067-4688 AG5067-4721 AG5067-4722 AG5067-4723 AG5067-4724 AG5067-4735 AG5067-4744 AG5067-4745 AG5067-4746 AG5067-4748 AG5067-5104 AG5067-5107 AG5067-5108 AG5067-5111 AG5067-5113 AG5067-5416 AG5067-6129 AG5500-1199 AG5500-1200 AG5500-1201 AG5500-1202 AG5500-1203 AG5500-1238 AG5500-1240 AG5500-1242 AG5500-1245 AG5500-1249 AG5500-1250 AG5500-1251 AG5500-1252 AG5500-1253 AG5500-1272 AG79841-87610 AGG1311-81600 AGG1311-81601 AGG1312-67300 AGG1312-67301 AGG1312-67500 AGG1312-87300 AGG1312-87305 AGG1312-87306 AGG1313-87304 AGG1313-87307 AGG1315-87303 AGG1315-87312 AGG1316-27301 AGG1316-87305 AGG1316-87309 AGG1316-87312 AGG1316-87315 AGG1316-87316 AGG1316-87317 AGG1316-87318 AGG1316-87319 AGG1316-87323 AGG1316-87328 AGG1328-87600 AGG1367-87304 AGG1375-87317 AGG4220-87000 AGG4226-60301 AGG5667-81001 AGG5667-81003 AGG5667-81004 AGG5667-81005 AG5022-6510 AG5023-2254 AG5065-9927 AG5065-9930 AG5065-9979 AG5065-9980 AG5067-1570 AG5067-1571 AG5067-4604 AG5067-4606 AG5067-4607 AG5067-4608 AG5067-4609 AG5067-4684 AG5067-4686 AG5067-4687 AG5067-4689 AG5067-4719 AG5067-4737 AG5067-4783 AG5067-5106 AG5067-5109 AG5067-5110 AG5067-5112 AG5067-5120 AG5067-5749 AG5067-5750 AG5067-5926 AG5067-6188 AG5067-6645 AG5067-6646 AG5067-6647 AG5067-6821 AG5500-1204 AG5500-1209 AG5500-1210 AG5500-1264 AG5500-1270 AG5500-1286 AG5500-1301 AG5500-1305 AG5500-1306 AG5500-1308 AG5500-1315 AG5500-1319 AG5500-1346 AG5500-1347 AG5500-1376 AG79826-87606 AG79826-87607 AG79835-87638 AG79841-87609 AG79846-87605 AGG1313-87308 AGG1315-87307 AGG1316-87304 AGG1316-87306 AGG1316-87313 AGG1316-87314 AGG1316-87326 AGG1361-67300 AGG1361-67301 AGG1361-67302 AGG1367-60301 AGG1367-87302 AGG1375-87304 AGG1375-87306 AGG1375-87311 AGG1375-87312 AGG1375-87318 AGG1375-87320 AGG2258-87301 AGG2260-87300 AGG2260-87301 AGG5611-60500 AGG5611-60501 AGG5611-60502 AGG5611-60503 AGG5667-81000 AGG5667-81002 AGG5667-81007 AG5004-0016 AG5004-0017 AG5004-0018 AG5004-0019 AG5004-0041 AG5022-6508 AG5022-6509 AG5061-3361 AG5065-4493 AG5065-9906 AG5065-9926 AG5067-1572 AG5067-4605 AG5067-4784 AG5067-5709 AG5067-5713 AG5067-5939 AG5067-5941 AG5067-6643 AG5067-6644 AG5067-6820 AG5500-1278 AG5500-1280 AG5500-1284 AG5500-1300 AG5500-1302 AG5500-1303 AG5500-1309 AG5500-1314 AG5500-1316 AG5500-1317 AG5500-1318 AG5500-1320 AG5500-1329 AG5500-1330 AG5500-1333 AG5500-1334 AG5500-1335 AG5500-1336 AG5500-1384 AG5500-1385 AG5500-1386 AG5500-1387 AG5500-1388 AG5500-1390 AG5500-1391 AG5500-1393 AG5500-1398 AG5500-1406 AG5500-1407 AG5500-1408 AG5500-1415 AG5500-1423 AG5500-1432 AG5500-1433 AG5500-1434 AG5500-1438 AG5500-1441 AG5500-1442 AG5500-1444 AG5500-1445 AG5500-1447 AG5500-1451 AG5500-1456 AG5500-1462 AG79826-87601 AG79826-87605 AG79826-87608 AGG1313-87302 AGG1313-87306 AGG1315-68708 AGG1315-87305 AGG1315-87318 AGG1315-87323 AGG1315-87328 AGG1315-87333 AGG1315-87338 AGG1316-87327 AGG1361-67304 AGG1361-67305 AGG1361-67306 AGG1361-67500 AGG1361-67501 AGG1373-87300 AGG1375-87300 AGG1375-87301 AGG1375-87302 AGG1375-87303 AGG1375-87305 AGG1375-87308 AGG1375-87309 AGG1375-87310 AGG1375-87313 AGG1375-87314 AGG1375-87316 AGG1375-87319 AGG1375-87321 AGG1375-87322 AGG1375-87323 AGG1375-87324 AGG1375-87325 AGG1375-87327 AGG1375-87400 AGG1377-87301 AGG4240-87300 AGG4240-87301 AGG4240-87302 AGG4240-87303 AGG4240-87304 AGG4240-87310 AGG4280-81300 AGG4280-81301 AGG4301-60056 AGG5611-67300 AGG5611-67301 AGG5615-87302 AGG7132-60002
Capillaries for HPLC, Agilent Technologies
Tubing Chromatography Tubing
High-quality LC capillaries in a large variety of materials and dimensions.
  • A variety of top-quality materials available such as stainless steel, titanium, PEEK-sleeved stainless steel, and PEEK-coated fused silica
  • Agilent 0.6 mm od flexible 316 L grade stainless steel capillaries are corrosion resistant and much easier to handle than conventional rigid 1.6 mm od capillaries
  • Color-coded for easy identification of inner diameters
  • Bio-inert titanium or PEEK-sleeved stainless steel capillaries that fit perfectly for bio applications
All Agilent LCMore Product Information