CrossLab Flow Cells for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
79883-68703 G1314-65050 5500-1342 5500-1343 5500-1344 5500-1345 G1314-60024 G1314-60025 G1314-60189 G1315-67305 G1315-80001 G1315-87101 G7115-60001 G7115-60002
AG79883-68703EA 268 USD
AG79883-68703 AGG1314-65050 AG5500-1342 AG5500-1343 AG5500-1344 AG5500-1345 AGG1314-60024 AGG1314-60025 AGG1314-60189 AGG1315-67305 AGG1315-80001 AGG1315-80002 AGG1315-87101 AGG7115-60001 AGG7115-60002
CrossLab Flow Cells for HPLC, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
High-quality flow cells for Agilent HPLC detectors to allow the best performance in all applications.
  • Designed for highest chromatographic performance
  • Flow cells for nano flow applications
Agilent provides a comprehensive range of high-quality flow cells for Agilent HPLC detectors to allow the best performance in all applications. Available solutions address the requirements of protein research applications or assays to assess the integrity of bio-therapeutic compounds. Bio-inert flow cells are available for Agilent multiple wavelength and diode arrayMore Product Information