CrossLab Graphite/Vespel Ferrules for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5062-3508 5181-3323 5062-3512 5062-3514 0100-1332 5062-3538 5062-3580 5062-3581 5181-3308 500-1004 CR213200 5181-3388 CP4784 5188-5314 5188-5315
AG5062-3508PK 115.3 USD
AG5062-3508 AG5181-3323 AG5062-3512 AG5062-3514 AG0100-1332 AG5062-3538 AG5062-3580 AG5062-3581 AG5181-3308 AG500-1004 AGCR213200 AG5181-3388 AGCP4784 AG5188-5314 AG5188-5315
CrossLab Graphite/Vespel Ferrules for GC
Fittings and Connectors
Graphite/Vespel (15/85%) ferrules, available in different configurations, provide leak-free connections between the column and injector on your GC.
  • Composed of 85% Vespel polyimide and 15% graphite
  • Non-porous material makes them ideal for MS applications
  • Withstand temperatures as high as 350 °C
  • Can be reused and reinstalled due their resistance to deformation
These ferrules are ideal for MS applications as their nonporous material prohibits oxygen contamination, and they can withstand temperatures as high as 350 °C. They are also widely used in GCMore Product Information