GC Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5181-1267 5181-3354 5181-3360 5181-8810 G4513-80206 G4513-80209 5181-1273 5181-3361 5182-0836 5182-0875 5183-4730 8010-0163 8010-0265 8010-0357 9301-0725 G4513-80203 G4513-80204 G4513-80208 G4513-80213 G4513-80220 G4513-80226 5181-3321 5181-8809 5182-0835 5183-0316 5183-2042 5183-4728 5183-4729 5188-5246 5188-5247 8004-0001 8010-0164 8010-0351 8010-0364 G4513-80200 G4513-80201 G4513-80202 G4513-80205 G4513-80215 G4513-80216 G4513-80229 G4513-80234 G4513-80235 G4513-80243 G4513-80244 G6500-80109 G6500-80116 5181-3356 5181-8806 5181-8813 5182-0834 5182-9799 5183-0314 8010-0118 8010-0129 8010-0130 8010-0168 8010-0174 8010-0270 8010-0352 8010-0355 8010-0356 8010-0358 8010-0363 8010-0371 9301-0714 G4200-80119 G4513-60560 G4513-60561 G4513-80210 G4513-80211 G4513-80212 G4513-80218 G4513-80221 G4513-80222 G4513-80241 G4513-80242 G6500-80107 G6500-80114 G6500-80115 5182-9622 5182-9626 5182-9734 5188-6485 5190-3984 8010-0090 8010-0138 8010-0169 8010-0173 8010-0184 8010-0197 8010-0266 8010-0271 8010-0353 8010-0360 8010-0361 8010-0362 G4200-80118 G4513-80207 G4513-80219 G4513-80223 G4513-80224 G4513-80228 G4513-80230 G4513-80233 G6500-80111 G6500-80113
AG5181-1267EA 72 USD
AG5181-1267 AG5181-3354 AG5181-3360 AG5181-8810 AGG4513-80206 AGG4513-80209 AG5181-1273 AG5181-3361 AG5182-0836 AG5182-0875 AG5183-4730 AG8010-0163 AG8010-0265 AG8010-0357 AG9301-0725 AGG4513-80203 AGG4513-80204 AGG4513-80208 AGG4513-80213 AGG4513-80220 AGG4513-80226 AG5181-3321 AG5181-8809 AG5182-0835 AG5183-0316 AG5183-2042 AG5183-4728 AG5183-4729 AG5188-5246 AG5188-5247 AG8004-0001 AG8010-0164 AG8010-0351 AG8010-0364 AGG4513-80200 AGG4513-80201 AGG4513-80202 AGG4513-80205 AGG4513-80215 AGG4513-80216 AGG4513-80229 AGG4513-80234 AGG4513-80235 AGG4513-80243 AGG4513-80244 AGG6500-80109 AGG6500-80116 AG5181-3356 AG5181-8806 AG5181-8813 AG5182-0834 AG5182-9799 AG5183-0314 AG8010-0118 AG8010-0129 AG8010-0130 AG8010-0168 AG8010-0174 AG8010-0270 AG8010-0352 AG8010-0355 AG8010-0356 AG8010-0358 AG8010-0363 AG8010-0371 AG9301-0714 AGG4200-80119 AGG4513-60560 AGG4513-60561 AGG4513-80210 AGG4513-80211 AGG4513-80212 AGG4513-80218 AGG4513-80221 AGG4513-80222 AGG4513-80241 AGG4513-80242 AGG6500-80107 AGG6500-80114 AGG6500-80115 AG5182-9622 AG5182-9626 AG5182-9734 AG5188-6485 AG5190-3984 AG8010-0090 AG8010-0138 AG8010-0169 AG8010-0173 AG8010-0184 AG8010-0197 AG8010-0266 AG8010-0271 AG8010-0353 AG8010-0360 AG8010-0361 AG8010-0362 AGG4200-80118 AGG4513-80207 AGG4513-80219 AGG4513-80223 AGG4513-80224 AGG4513-80228 AGG4513-80230 AGG4513-80233 AGG6500-80111 AGG6500-80113
GC Autosampler Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Agilent syringes are what you need for precise, effective, sample delivery from your GC autosampler.
  • Blue line syringes precisely match with your autosampler’s stroking mechanism for more accurate volume delivery
  • The smooth needle of the blue line syringes reduces septum coring and keeps your system working at its full potential
  • Blue line syringes are available in a choice of PTFE-tipped or metal fitted plunger to meet your application needs
  • Blue line syringes are delivered in easy-open, environmentally friendly packaging
  • Lot numbers are printed directly
More Product Information