Capillaries for Shimadzu LC Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
8001-0807 8001-0821 8001-0822 8001-0823 8001-0824
AG8001-0807EA 52.2 USD
AG8001-0807 AG8001-0821 AG8001-0822 AG8001-0823 AG8001-0824 AG8001-8020
Capillaries for Shimadzu LC Systems, Agilent Technologies
Tubing Chromatography Tubing
Capillaries and tubing suitable for most standard applications on selected Shimadzu HPLC systems.
  • Made of ultra-clean stainless steel tubing
  • Ideal for most standard applications, and the best choice for reliable high-pressure sealing
  • Smooth inner surface for the lowest backpressure
  • Precision cutting for burr-free, square-cut ends without inner-diameter distortion
  • Both ends accept standard fittings
  • Predefined lengths for specific path locations to avoid peak broadening
  • Individually tested to ensure reproducibility
  • Available in non pre-swaged