VersaPlate Well Plate Tubes
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7550301C 75503025 7550401C 75504050 7550501C 7550601C 7550701C 75507025 7550801C 75508050 75509050 7551101C 7551201C 7551301C 7551401C 7551501C 7552003C 75520060 7552201C 7552301C 7552401C 7552501C 7553502C 75580030 75590030
AG7550301CPK 319 USD
AG7550301C AG75503025 AG7550401C AG75504050 AG7550501C AG7550601C AG7550701C AG75507025 AG7550801C AG75508050 AG75509050 AG7551101C AG7551201C AG7551301C AG7551401C AG7551501C AG7552003C AG75520060 AG7552201C AG7552301C AG7552401C AG7552501C AG7553502C AG75580030 AG75590030 AG75590060
VersaPlate Well Plate Tubes
Tubes Test Tubes
VersaPlate accelerates solid phase extraction (SPE) and solid supported liquid extraction (SLE) method development in a 96-well plate format with removable tubes. Configurations can be customized depending on the number of samples.
  • Choose the level of automation that best suits your needs while you use the 96-well plate format or create your own customized plates
  • Develop an SPE method faster, using liquid handling automation or multichannel pipettes, or use individual tubes to quickly work out method conditions and transfer to 96-well processing
  • Use only
More Product Information