CrossLab Tubing, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5180-4196 5021-7107 1300502506 7157-0210 CP4006 CP4008 CP4025 CP4260 5062-2419 5190-6983 AL3000 AL3002 AL3003 CP4003 CP4005 CP4013 CP4024 CP4026 CP4032 CP4049 CP4056 R000383005
AG5180-4196EA 158.2 USD
AG5180-4196 AG5021-7107 AG1300502506 AG7157-0210 AGCP4006 AGCP4008 AGCP4025 AGCP4260 AG5062-2419 AG5190-6983 AGAL3000 AGAL3002 AGAL3003 AGCP4003 AGCP4005 AGCP4013 AGCP4024 AGCP4026 AGCP4032 AGCP4049 AGCP4056 AGR000383005
CrossLab Tubing, Agilent Technologies
Tubing Chromatography Tubing
Copper, stainless steel, and UltiMetal Plus tubing for GC applications.
  • Copper tubing can be used for most applications, except those requiring the highest purity gas delivery system
  • Stainless steel and UltiMetal Plus tubing are appropriate for more demanding applications
Agilent GC tubing includes high-quality copper, stainless steel, and UltiMetal Plus tubing, which are manufactured and cleaned for chromatographic use. Agilent tubing comes in different lengths and depending on your flow requirements, is available in ¹/₁₆, ¹/₈, or ¹/₄". UltiMetalMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable