Autosampler Needles and Plungers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G1313-87201 G1313-87202 G7129-87200 G7129-87201 G1313-87203 G7129-87202 5190-1496 5190-1548 5190-1549 G1329-80001 G3160-65306 G4277-80125 RF0050T 5181-7442 5181-8811 5182-0830 5190-1461 5190-1473 5190-1477 5190-1478 5190-1482 5190-1538 5190-1550 5190-1571 5190-1581 5190-1582 8001-0005 8001-0007 8001-0014 8002-0005 8010-0367 G1811-23200 RF0050T-S RF0052T
AGG1313-87201EA 98.95 USD
AGG1313-87201 AGG1313-87202 AGG7129-87200 AGG7129-87201 AGG1313-87203 AGG7129-87202 AG5190-1496 AG5190-1548 AG5190-1549 AGG1329-80001 AGG3160-65306 AGG4277-80125 AGRF0050T AG5181-7442 AG5181-8811 AG5182-0830 AG5190-1461 AG5190-1473 AG5190-1477 AG5190-1478 AG5190-1482 AG5190-1538 AG5190-1550 AG5190-1571 AG5190-1581 AG5190-1582 AG8001-0005 AG8001-0007 AG8001-0014 AG8002-0005 AG8010-0367 AGG1811-23200 AGRF0050T-S AGRF0052T AGRF0052T-S
Autosampler Needles and Plungers, Agilent Technologies
Syringe Needles
Not ready for a new syringe? Try out our replacement needles and plungers.
  • Available for a variety of syringe makes in a choice of PTFE-tipped or metal fitted plunger to meet your application needs
  • Environmentally friendly packaging, with an improved design that reduces waste
  • Fresh needles help to reduce septum coring and keeps your system working at its full potential
  • Help to maintain a chemically inert fluid path of stainless steel, borosilicate Type I glass, or PTFE
Agilent replacement needles and plungers ensure your syringes continue to functionMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable