CrossLab Capillary and Tubing Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5065-9939 5061-3304 5065-9937 5067-6614 5067-6615 5067-6707 5004-0003 5013-0002 5061-3315 5065-9938 5067-4234 5067-4249 5067-4250 5067-4251 5067-4252 5067-4270 5067-4294 5067-4300 5067-4633 5067-6181 5067-6187 5067-6596 5067-6597 5067-6598 5067-6697 5067-6708 5067-6800 5067-6802 G1316-68716 G5611-68711 5067-6801 5067-6804
AG5065-9939KT 1090.7 USD
AG5065-9939 AG5061-3304 AG5065-9937 AG5067-6614 AG5067-6615 76537-448 AG5004-0003 AG5013-0002 AG5061-3315 AG5065-9938 AG5067-4234 AG5067-4249 AG5067-4250 AG5067-4251 AG5067-4252 AG5067-4270 AG5067-4294 AG5067-4300 AG5067-4633 AG5067-6181 AG5067-6187 AG5067-6596 AG5067-6597 AG5067-6598 AG5067-6697 AG5067-6708 AG5067-6800 AG5067-6802 AGG1316-68716 AGG5611-68711 AG5067-6801 AG5067-6804
CrossLab Capillary and Tubing Kits for HPLC, Agilent Technologies
Tubing Chromatography Tubing
A complete set of system capillaries and fittings to efficiently ramp up usage of LC instruments.
  • Carefully selected capillary dimensions (inner diameter and length) to minimize extra-column volume and optimize separation efficiency
  • Peace of mind of having everything needed to make necessary connections on your system
  • InfinityLab Quick Connect and Quick Turn fittings in the InfinityLab capillary kits ensure ease of use and zero dead-volume column connection
Agilent InfinityLab capillary kits are complete sets of system capillaries, includingMore Product Information