Capillary Kits for HPLC
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5023-2255 5067-4601 5023-2256 5023-2257 5023-2258 5065-4495 5065-9972 5067-4248 5067-4730 5067-4767 5067-4769 5067-5708 5067-6621 5067-6723 5067-6724 5067-6867 5067-6868 5067-6869 5067-6870 G4309-68715 5067-4800 5067-5103
AG5023-2255KT 273 USD
AG5023-2255 AG5067-4601 AG5023-2256 AG5023-2257 AG5023-2258 AG5065-4495 AG5065-9972 AG5067-4248 AG5067-4730 AG5067-4767 AG5067-4769 AG5067-5708 AG5067-6621 AG5067-6723 AG5067-6724 AG5067-6867 AG5067-6868 AG5067-6869 AG5067-6870 AGG4309-68715 AG5067-4800 AG5067-5103
Capillary Kits for HPLC
Tubing Chromatography Tubing
System capillaries and fittings for efficienct ramp up of usage of your LC instruments.
  • Carefully selected capillaries dimensions (inner diameter and length) to minimize extra-column volume and optimize separation efficiency
  • Get peace of mind from having everything at hand for all necessary connections on your new systems
  • InfinityLab Quick Connect and Quick Turn fittings in the InfinityLab capillary kits ensure ease-of-use and zero-dead-volume column connections
Agilent InfinityLab capillary kits include a complete set of system capillaries, includingMore Product Information