InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5043-9300 5043-9310 5043-9302 5043-9305 5043-9311 5043-9312 5065-4402 5043-9308 5043-9309 5043-9313 5043-9314 5043-9315
AG5043-9300PK 322.9 USD
AG5043-9300 AG5043-9310 AG5043-9302 AG5043-9305 AG5043-9311 AG5043-9312 AG5065-4402 AG5043-9308 AG5043-9309 AG5043-9313 AG5043-9314 AG5043-9315
InfinityLab Well Plates, Agilent Technologies
Microplates Deep Well Microplates
A comprehensive selection of InfinityLab well plates and sealing mats for the HPLC workflow.
  • Ensure a perfect fit between well plates and slitted sealing mats
  • Access the right formats to ensure each well plate sampler setup is perfectly equipped for all HPLC applications
  • Optimized for high performance in your chromatography workflow
  • A large range of well volumes and package quantities provide the flexibility needed to suit your lab size and your needs
  • Compliant with worldwide ANSI and SLAS standards
Agilent provides a comprehensive selection ofMore Product Information