CrossLab Macrocells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
1000-0544 5061-3387 6610000800 6610016100 5061-3384 5061-3385 5061-3386 5063-6549 6610001100 6610008800 6610016000 5062-2477 5063-6547 5063-6550 5063-6551 5063-6553 5063-6557 6610016200 6610016300 6610016400
AG1000-0544PK 477.8 USD
AG1000-0544 AG5061-3387 AG6610000800 AG6610016100 AG5061-3384 AG5061-3385 AG5061-3386 AG5063-6549 AG5063-6552 AG5063-6554 AG6610001100 AG6610008800 AG6610016000 AG5062-2477 AG5063-6546 AG5063-6547 AG5063-6548 AG5063-6550 AG5063-6551 AG5063-6553 AG5063-6557 AG6610016200 AG6610016300 AG6610016400
CrossLab Macrocells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Agilent offers a wide range of rectangular cuvettes with optical path lengths from 1 to 100 mm.
  • Tried and tested over decades, used in countless applications
  • Exceptional accuracy in terms of optical path length and parallelism
  • Unique surface flatness of the optical windows
  • High temperature stability
  • High chemical resistance
  • Outstanding measurement reproducibility
  • Guaranteed transmission of >80% from 200 - 3500 nm (depending on the cell window material)
  • Cells have either an open top (with a fitted lid) or a fused cover which is sealed using a PTFE
More Product Information