CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
17-4000 17-4001 17-4003 17-4005 17-4010 17-4011 17-4020 17-4021 17-4040 17-4045 17-4100 17-4140 17-4004 17-4015 17-4025 17-4050 17-4055 17-4110 17-4120 17-4130 K1200-00855
AG17-4000PK 179 USD
AG17-4000 AG17-4001 AG17-4003 AG17-4005 AG17-4010 AG17-4011 AG17-4020 AG17-4021 AG17-4040 AG17-4045 AG17-4100 AG17-4140 AG17-4004 AG17-4015 AG17-4025 AG17-4050 AG17-4055 AG17-4110 AG17-4120 AG17-4130 AGK1200-00855
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Dissolution Apparatus
Made of either color-coded UHMWPE or PVDF, the full flow filter prevents clogging.
  • Available in ultrahigh molecular weight polyethylene (UHMWPE) or polyvinylidene fluoride (PVDF)
  • Full flow filter offers increased surface area to prevent clogging
  • Low protein-binding PVDF filter material available
  • Usable with ¹/₈" diameter cannulas
  • Available in packs of 100 or 1000
  • Full flow filters are color coded for easy identification
  • Certificates of analysis can be provided on request
  • Standard conventional filter tips are made of polyethylene fluoride and can also be
More Product Information