CrossLab Dissolution Shafts, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
14-3613 14-3631 14-3636 16-3631 13-3614 13-3615 13-3622 13-3629 13-3630 13-3632 14-3612 14-3620 14-3624 14-3634 16-3613
AG14-3613EA 270 USD
AG14-3613 AG14-3631 AG14-3636 AG16-3631 AG13-3614 AG13-3615 AG13-3622 AG13-3629 AG13-3630 AG13-3632 AG14-3612 AG14-3620 AG14-3624 AG14-3634 AG16-3613
CrossLab Dissolution Shafts, Agilent Technologies
Dissolution Apparatus
Shafts of different materials and lengths, individually serialized to meet standard USP design.
  • Electropolished stainless steel, PTFE-coated, gold-plated or mini basket shafts available
  • Individually serialized
  • Three-clip shafts to meet standard USP design
  • O-ring shafts for automated systems
  • Conical shafts to support air bubble dispersion
  • A range of basket shaft lengths are available to accommodate various models and volume configurations
Agilent provides stainless steel, PTFE-coated, gold-plated, or mini shafts that are individually serialized.More Product Information