Dissolution Evaporation Covers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
12-6300 12-6305 12-6310 12-6311 12-6315 12-6320 12-6327 12-6329 12-6330 27-1500 27-1510
AG12-6300EA 125.2 USD
AG12-6300 AG12-6305 AG12-6310 AG12-6311 AG12-6315 AG12-6320 AG12-6327 AG12-6328 AG12-6329 AG12-6330 AG27-1500 AG27-1510
Dissolution Evaporation Covers, Agilent Technologies
Dissolution Apparatus
A variety of evaporation covers are available to fit your apparatus and vessels.
  • The evaporation covers automatically travel with the apparatus driveunit using either the alignment posts or DDM assemblies
  • The standard evaporation cover on the 708-DS is designed to allow less than 1% evaporative loss over a 24-hour period under normal operating conditions
  • Agilent also offers other low loss covers that can reduce evaporative loss even further
A variety of evaporation covers are available to fit your apparatus and vessels. Evaporation plugs are used toMore Product Information