Propidium Iodide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide

J66764-MC
AAJ66764-MCEA 195 USD
AAJ66764-MC
Propidium Iodide
Propidium iodide

Propidium iodide


CAS No.: 25535-16-4 Molecular Formula: C27H34I2N4 Formula Weight: 668.39 Physical Form: Solid Sensitivity: Light Sensitive; Hygroscopic Appearance: Dark red Melting Point: 220-225° Solubility: Soluble in water at 1mg/ml Beilstein Reference: 3843838 MDL No.: MFCD00011921

Formula: C₂₇H₃₄I₂N₄
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00011921
CAS Number: 25535-16-4
EINECS: 247-081-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR