Laemmli SDS sample buffer, reducing (6X), Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61337-AC J61337-AD
AAJ61337-ACEA 186.37 USD
AAJ61337-AC AAJ61337-AD
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Buffers Electrophoresis Buffers Loading Buffers

Liquid


Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR