TRIS, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61141-AP J61141-K2 J61144-AP J61144-K2 J62131-AP J62131-K2 J62287-AP J62287-K2 J63735-AP J63735-K2
AAJ61009-APEA 88.91 USD
AAJ61009-AP AAJ61009-K2 AAJ61141-AP AAJ61141-K2 AAJ61144-AP AAJ61144-K2 AAJ61213-AP AAJ61213-K2 AAJ61537-AP AAJ61537-K2 AAJ61787-AP AAJ61787-K2 AAJ61944-AP AAJ61944-K2 AAJ62131-AP AAJ62131-K2 AAJ62287-AP AAJ62287-K2 AAJ62551-AP AAJ62551-K2 AAJ62552-AP AAJ62552-K2 AAJ62736-AP AAJ62736-K2 AAJ62778-AP AAJ62778-K2 AAJ62944-AP AAJ62944-K2 AAJ63134-AP AAJ63134-K2 AAJ63735-AP AAJ63735-K2
TRIS, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Tris (hydroxymethyl) aminomethane.


CAS No.: 77-86-1
MDL No.: MFCD00004679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR