PBDB-T-2F (PCE14), OPV - Organic Photovoltaics

Supplier: Thermo Scientific Chemicals

Synonyms: PCE135

H66179-MC H66179-MD H66179-ME
AAH66179-MCEA 583 USD
AAH66179-MC AAH66179-MD AAH66179-ME
PBDB-T-2F (PCE14), OPV - Organic Photovoltaics
PBDB-T-2F (PCE14)
Formula: (C₆₈H₇₆F₂O₂S₈)n
CAS Number: 1802013-83-7

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.85 ± 0.02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR