2,2-Diphenylglycine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: alpha,alpha-Diphenylglycine, Diphenylglycin, 2-Amino-2,2-diphenylacetic acid

B22650-14 B22650-06 B22650-22
AAB22650-14EA 341 USD
AAB22650-14 AAB22650-06 AAB22650-22
2,2-Diphenylglycine 98%
2,2-Diphenylglycine

Purity: 98% CAS No.: 3060-50-2 Molecular Formula: C14H13NO2 Formula Weight: 227.26 Melting Point: ca 246° dec. MDL No.: MFCD00008048

Formula: C₁₄H₁₃NO₂
Melting Pt: ca 246 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008048
CAS Number: 3060-50-2
EINECS: 221-299-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR