Boranepyridine complex 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine Borane, Borane, pyridine complex

L13178-09 L13178-18
AAAL13178-09EA 57.6 USD
AAAL13178-09 AAAL13178-18
Boranepyridine complex 95%
Boranepyridine complex

Fieser: 8,50 9,59 11,69 12,65 18,58 19,43 21,65


Purity: 95% CAS No.: 110-51-0 Molecular Formula: C5H8BN Formula Weight: 92.93 Storage Temperature: 2°C to 8°C Flash Point: 73°(163°F) Refractive Index: 1.5320 Odor: Unpleasant Melting Point: 9-11° Boiling Point: 65°/1mm Beilstein Reference: 3674913 UN No.: 2810 MDL No.: MFCD00012435

Formula: C₅H₈BN
Boiling Pt: 65 °C/1 mm
Melting Pt: 9…11 °C
Density: 0.920
Flash Pt: 73 °C(163 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00012435
CAS Number: 110-51-0
EINECS: 203-773-4
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR