Allylthiourea 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea

L03377-14
AAAL03377-14EA 38.2 USD
AAAL03377-14
Allylthiourea 98%
Allylthiourea

Purity: 98% CAS No.: 109-57-9 Molecular Formula: C4H8N2S Formula Weight: 116.19 Odor: Unpleasant Melting Point: 70-78° Beilstein Reference: 1071470 Merck Reference: 14,9362 UN No.: 2811 MDL No.: MFCD00004940

Formula: H₂CCHCH₂NHC(S)NH₂
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: 70…78 °C
Density: 1.220
Storage Temperature: Ambient
MDL Number: MFCD00004940
CAS Number: 109-57-9
EINECS: 203-683-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,9362

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR