4-Nitro-2-(trifluoromethyl)aniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-nitro-α,α,α-trifluortoluene, 2-Amino-5-nitro-α,α,α-trifluorotoluene, 4-Nitro-2-trifluoromethylaniline, 2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine

L00977-06 L00977-14
AAAL00977-06EA 32.2 USD
AAAL00977-06 AAAL00977-14
4-Nitro-2-(trifluoromethyl)aniline 98%
4-Nitro-2-(trifluoromethyl)aniline

Purity: 98% CAS No.: 121-01-7 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 90-93° Beilstein Reference: 2121347 UN No.: 2811 MDL No.: MFCD00007365

Formula: C₇H₅F₃N₂O₂
MW: 206.12 g/mol
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
EINECS: 204-443-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR