7-Azaindole-5-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Azaindole-5-boronic Acid Pinacol Ester

H50044-MD H50044-03
AAAH50044-MDEA 46.2 USD
AAAH50044-MD AAAH50044-03
7-Azaindole-5-boronic acid pinacol ester 97%
7-Azaindole-5-boronic acid pinacol ester

Purity: 97% CAS No.: 754214-56-7 Molecular Formula: C13H17BN2O2 Formula Weight: 244.10 Melting Point: 241-245° MDL No.: MFCD08060937

Formula: C₁₃H₁₇BN₂O₂
MW: 244.101 g/mol
Melting Pt: 241…245 °C
Storage Temperature: Ambient
MDL Number: MFCD08060937
CAS Number: 754214-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR