Congo red indicator

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate), CI 22120, Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate), Direct Red 28

B24310-22 B24310-36 B24310-14
AAAB24310-22EA 163.64 USD
AAAB24310-22 AAAB24310-36 AAAB24310-14
Congo red indicator
Congo Red

CAS No.: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Formula Weight: 696.67 Odor: Odorless Melting Point: 360° Beilstein Reference: 3894858 Merck Reference: 14,2498 MDL No.: MFCD00004028

Formula: C₃₂H₂₂N₆Na₂O₆S₂
Melting Pt: >360 °C
Storage Temperature: Ambient
MDL Number: MFCD00004028
CAS Number: 573-58-0
EINECS: 209-358-4
Merck Index: 14,2498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR