Sodium trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trifluoromethanesulphonic acid sodium salt, Trifluoromethanesulfonic acid sodium salt

B20776-06 B20776-22 B20776-14
AAAB20776-06EA 58.2 USD
AAAB20776-06 AAAB20776-22 AAAB20776-14
Sodium trifluoromethanesulfonate 98%
Sodium trifluoromethanesulphonate

Purity: 98% CAS No.: 2926-30-9 Molecular Formula: CF3SO3Na Formula Weight: 172.06 Melting Point: 251-255° Beilstein Reference: 3728797

Formula: CF₃NaO₃S
Melting Pt: 251…255 °C
Storage Temperature: Ambient
MDL Number: MFCD00061607
CAS Number: 2926-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR