4'-Iodoacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone

A16794-22 A16794-06 A16794-14
AAAA16794-22EA 334 USD
AAAA16794-22 AAA16794-06 AAA16794-14
4'-Iodoacetophenone 98%
4'-Iodoacetophenone

Purity: 98% CAS No.: 13329-40-3 Molecular Formula: C8H7IO Formula Weight: 246.05 Melting Point: 82-84° Beilstein Reference: 1857412

Formula: C₈H₇IO
MW: 246.05 g/mol
Boiling Pt: 145…150 °C (20 mmHg)
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR