Copper(II) acetylacetonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

A14920-0B A14920-22 A14920-18
AAAA14920-0BEA 464 USD
AAAA14920-0B AAAA14920-14 AAAA14920-22 AAAA14920-18 AAAA14920-36
Copper(II) acetylacetonate 98%
Copper (II) acetylacetonate

Purity: 98% CAS No.: 13395-16-9 Molecular Formula: C10H14CuO4 Formula Weight: 261.76 Odor: Weak Melting Point: ca 286° dec. Boiling Point: 160°/10mm Beilstein Reference: 4157957 MDL No.: MFCD00000016

Formula: C₁₀H₁₄CuO₄
Boiling Pt: 160 °C/10 mm
Melting Pt: ∼ 286 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000016
CAS Number: 13395-16-9
EINECS: 236-477-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR