Urea 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

A12360-0C A12360-36 A12360-0E
AAAA12360-0CEA 210.69 USD
AAAA12360-0C AAAA12360-36 AAAA12360-0E
Urea 98+%
Urea

Used as a protein denaturant


Purity: 98+% CAS No.: 57-13-6 Molecular Formula: NH2CONH2 Formula Weight: 60.06 Odor: Ammonia-like pH: 9,2-9,5 Melting Point: 130-135° Beilstein Reference: 635724 Merck Reference: 14,9867 MDL No.: MFCD00008022

Formula: NH₂CONH₂
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 130…135 °C
Density: 1.335
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 14,9867
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR