Hydrocortisone 98%

Supplier: Thermo Scientific Chemicals
Danger

A16292-06 A16292-03
AAA16292-06EA 158 USD
AAA16292-06 AAA16292-03
Hydrocortisone 98%
Hydrocortisone

A natural corticosteroid and anti-inflammatory compound


Purity: 98% CAS No.: 50-23-7 Molecular Formula: C21H30O5 Formula Weight: 362.47 Physical Form: Powder Appearance: White Odor: Odorless Melting Point: 213-215° Beilstein Reference: 1354819 Merck Reference: 14,4787 MDL No.: MFCD00011654


A natural corticosteroid and anti-inflammatory compound

Formula: C₂₁H₃₀O₅
MW: 362.47 g/mol
Boiling Pt: 567
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00011654
CAS Number: 50-23-7
EINECS: 200-020-1
Merck Index: 14,4787

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR