4-Bromoaniline 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15126-30 A15126-18 A15126-0B
AAA15126-30EA 126.46 USD
AAA15126-30 AAA15126-18 AAA15126-0B
4-Bromoaniline 98+%
4-Bromoaniline

Purity: 98+% CAS No.: 106-40-1 Molecular Formula: C6H6BrN Formula Weight: 172.03 Physical Form: Cryst solid Odor: Characteristic Boiling Point: 222-224° Beilstein Reference: 742031 Merck Reference: 14,1404 UN No.: 2811 MDL No.: MFCD00007822

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 223
Melting Pt: 60…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00007822
CAS Number: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1 III
Merck Index: 14,1404

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR