3,5-Dimethoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A15109-14 A15109-22 A15109-06
AAA15109-14EA 277 USD
AAA15109-14 AAA15109-22 AAA15109-06
3,5-Dimethoxybenzaldehyde 98%
3,5-Dimethoxybenzaldehyde

Purity: 98% CAS No.: 7311-34-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 44-48° Boiling Point: 115-118°/1mm Beilstein Reference: 2042374 MDL No.: MFCD00003366

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 277
Melting Pt: 48
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003366
CAS Number: 7311-34-4
EINECS: 230-772-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR