HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

A14777-09 A14777-18 A14777-30
AAA14777-09EA 28.65 USD
AAA14777-09 AAA14777-18 AAA14777-30
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 99%
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

A zwitterionic buffer


Purity: 99% CAS No.: 7365-45-9 Molecular Formula: C8H18N2O4S Formula Weight: 238.31 Physical Form: Powder Appearance: White Melting Point: ca 236° dec. Beilstein Reference: 883043 Merck Reference: 14,4654 MDL No.: MFCD00006158


A zwitterionic buffer

Formula: C₈H₁₈N₂O₄S
Melting Pt: ∼ 236 °C (dec.)
Density: 1.448 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 14,4654
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR