Glutaric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,5-Pentanedioic acid

A14595-22 A14595-14 A14595-36
AAA14595-22EA 83.72 USD
AAA14595-22 AAA14595-14 AAA14595-36
Glutaric acid 99%
Glutaric acid

Purity: 99% CAS No.: 110-94-1 Molecular Formula: C5H8O4 Formula Weight: 132.12 Melting Point: 95-99° Boiling Point: 200°/20mm Merck Reference: 14,4473 MDL No.: MFCD00004410

Formula: C₅H₈O₄
Boiling Pt: 200 °C/20 mm
Melting Pt: 95…99 °C
Density: 1.429
Storage Temperature: Ambient
MDL Number: MFCD00004410
CAS Number: 110-94-1
EINECS: 203-817-2
Merck Index: 14,4473

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR