Pyruvic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

A13875-0E A13875-22 A13875-36
AAA13875-0EEA 809 USD
AAA13875-0E AAA13875-22 AAA13875-36
Pyruvic acid 98%
Pyruvic acid

Purity: 98% CAS No.: 127-17-3 Molecular Formula: C3H4O3 Formula Weight: 88.06 Storage Temperature: 2°C to 8°C Flash Point: 83°(181°F) Physical Form: Liquid Density: 1.267 Refractive Index: 1.4295 Appearance: Colorless to yellow Odor: Pungent pH: 1.2 Melting Point: 11-12° Boiling Point: 164-166° Beilstein Reference: 506211 Merck Reference: 14,8021 UN No.: 3265 MDL No.: MFCD00002585

Formula: C₃H₄O₃
Boiling Pt: 164…166 °C
Melting Pt: 11…12 °C
Density: 1.267
Flash Pt: 83 °C (181 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,III
Merck Index: 14,8021

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR