1-Hexanesulfonic acid sodium salt 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate

A13278-14 A13278-06 A13278-22
AAA13278-14EA 238.79 USD
AAA13278-14 AAA13278-06 AAA13278-22
1-Hexanesulfonic acid sodium salt 99%
1-Hexanesulphonic acid sodium salt

Purity: 99% CAS No.: 2832-45-3 Molecular Formula: CH3(CH2)5SO3Na Formula Weight: 188.22 Physical Form: Powder Melting Point: 300° Beilstein Reference: 3727014 MDL No.: MFCD00007542

Formula: C₆H₁₃O₃SNa
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR