α,α'-Dichloro-o-xylene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Xylylene Dichloride, o-Xylylene chloride, α,α′-Dichloro-o-xylene, α,α'-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, Alpha,Alpha’-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, o-Xylene dichloride

A12500-18 A12500-30
AAA12500-18EA 77.36 USD
AAA12500-18 AAA12500-30
α,α'-Dichloro-o-xylene 98%
α,α'-Dichloro-o-xylene

Purity: 98% CAS No.: 612-12-4 Molecular Formula: C8H8Cl2 Formula Weight: 175.06 Flash Point: 107°(224°F) Odor: Pungent Boiling Point: 246° Beilstein Reference: 2043675 UN No.: 2928 MDL No.: MFCD00000903

Formula: C₈H₈Cl₂
Boiling Pt: 246 °C
Melting Pt: 53…57 °C
Density: 1.393 g/cm³ (0 °C)
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000903
CAS Number: 612-12-4
EINECS: 210-291-8
UN: 2928
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR