Mercury(II) acetate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

A12478-22 A12478-36 A12478-14
AAA12478-22EA 106.48 USD
AAA12478-22 AAA12478-36 AAA12478-14
Mercury(II) acetate 98+%
Mercury (II) acetate

Purity: 98+% CAS No.: 1600-27-7 Molecular Formula: C4H6HgO4 Formula Weight: 318.68 Melting Point: 179-182° Beilstein Reference: 3563831 Merck Reference: 14,5873 UN No.: 1629 MDL No.: MFCD00012165

Formula: (H₃CCOO)₂Hg
MW: 318.68 g/mol
Melting Pt: 179…182 °C
Density: 3.28
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1 II
Merck Index: 14,5873

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR