4-Isopropylbenzonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

A11954-03 A11954-06 A11954-14
AAA11954-03EA 43.2 USD
AAA11954-03 AAA11954-06 AAA11954-14
4-Isopropylbenzonitrile 95%
4-Isopropylbenzonitrile

MDL: MFCD00052966 Beilstein Registry No.: 1932883


Purity: 95% CAS No.: 13816-33-6 Molecular Formula: C10H11N Formula Weight: 145.21 Refractive Index: 1.5180 Boiling Point: 88-90°/5mm MDL No.: MFCD00052966

Formula: C₁₀H₁₁N
MW: 145.2
Boiling Pt: 88…90 °C (5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052966
CAS Number: 13816-33-6
EINECS: 237-492-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR