Diethylenetriamine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine

A11033-AP A11033-0F A11033-AE
AAA11033-APEA 44.77 USD
AAA11033-AP AAA11033-0F AAAA11033-AE
Diethylenetriamine 99%
Diethylenetriamine

MDL: MFCD00008171 Beilstein Registry No.: 605314


Purity: 99% CAS No.: 111-40-0 Molecular Formula: C4H13N3 Formula Weight: 103.17 Flash Point: 94°(201°F) Refractive Index: 1.4830 Odor: Ammonia-like Melting Point: -40° Boiling Point: 200-204° Beilstein Reference: 605314 UN No.: 2079 MDL No.: MFCD00008171

Formula: C₄H₁₃N₃
Boiling Pt: 200…204 °C
Melting Pt: –40 °C
Density: 0.952
Flash Pt: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008171
CAS Number: 111-40-0
EINECS: 203-865-4
UN: 2079
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR